Anti-Pseudomonas Mallei Mouse Monoclonal Antibody [clone: 3D11]

Supplier: GeneTex
GTX10011
89350-566EA 382.65 USD
89350-566
Anti-Pseudomonas Mallei Mouse Monoclonal Antibody [clone: 3D11]
Antibodies
Mouse monoclonal [3D11] to Pseudomonas mallei LPS

ELISA: Use at an assay dependent dilution. WB: Use at an assay dependent dilution. Not tested in other applications. Optimal dilutions/concentrations should be determined by the end user.

Type: Primary
Antigen:
Clonality: Monoclonal
Clone: 3D11
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR