Anti-PSA Mouse Monoclonal Antibody [clone: 8A6]

Supplier: GeneTex
GTX10187
89350-676EA 726.31 USD
89350-676
Anti-PSA Mouse Monoclonal Antibody [clone: 8A6]
Antibodies
Mouse monoclonal [8A6] to PSA

ELISA: Use at an assay dependent dilution. For the detection of free PSA use in ELISA as an antibody pair with either: GTX10185 coating and GTX10187 as conjugate, or GTX10184 coating and GTX10187 as conjugate, or GTX10187 coating and 10185 as conjugate. Not tested in other applications. Optimal dilutions/concentrations should be determined by the end user.

Type: Primary
Antigen: PSA
Clonality: Monoclonal
Clone: 8A6
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR