Anti-TFF1 Mouse Monoclonal Antibody [clone: pS2.1]

GTX23079
89351-852EA 376.78 USD
89351-852
Anti-TFF1 Mouse Monoclonal Antibody [clone: pS2.1]
Mouse monoclonal [pS2.1] to pS2

IHC(formalin fixed paraffin embedded): Use at a concentration of 2-4?g/ml for 30 minutes at RT. For immunohistochemistry, antigen retrieveal is not required. Not tested in other applications. Optimal dilutions/concentrations should be determined by the end user.

Type: Primary
Antigen: TFF1
Clonality: Monoclonal
Clone: pS2.1
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR