Anti-Kinase D2 Rabbit Polyclonal Antibody

Supplier: GeneTex
GTX78137
89366-514EA 481.52 USD
89366-514
Anti-Kinase D2 Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal to Protein Kinase D2 (PKD2)

Type: Primary
Antigen: Kinase D2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR