Anti-G Chicken Polyclonal Antibody

Supplier: GeneTex
GTX77599
89353-818EA 601.14 USD
89353-818
Anti-G Chicken Polyclonal Antibody
Antibodies
Type: Primary
Antigen: G
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Chicken
Isotype: IgY
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR