Anti-G Rabbit Polyclonal Antibody

Supplier: GeneTex
GTX27248
89362-518EA 788.34 USD
89362-518
Anti-G Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: G
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Other
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR