Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-C Mouse Monoclonal Antibody [clone: HLW-C]

Supplier: Genetex

GTX18702
89360-516EA 358.9 USD InStock
89360-516
Anti-C Mouse Monoclonal Antibody [clone: HLW-C]
Antibodies
Protein C is a vitamin K-dependent serine protease zymogen. Purified human activated protein C selectively destroys factors Va and VIII:C in human plasma and thus has an important anticoagulant role. Protein C deficiency has been associated with inherited thrombophilia. In its primary structure, protein C is similar to the prothrombin group of blood coagulation factors. It most closely resembles factor X and has light and heavy polypeptide chains linked by disulfide bridges.

Type: Primary
Antigen: C
Clonality: Monoclonal
Clone: HLW-C
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty