Anti-PRMT6 Rabbit Polyclonal Antibody

Supplier: GeneTex
GTX12192
89359-254EA 618.37 USD
89359-254
Anti-PRMT6 Rabbit Polyclonal Antibody
Antibodies
Postranslational modifications of proteins play an mportant role in the regulation of protein function, tability and localization. Such modifications occur on different amino acids and include phosphorylation,

IF: Use at a concentration of 3 - 4 ?g/ml (recommended using paraformaldehyde fixed HeLa cells). IP: 2.5 - 5 ?g of the antibody immunoprecipitates PRMT6 protein from extracts of PRMT6 transfected 293T cells. WB: Use at a concentration of 0.25 - 0.5 ?g/ml (recommended using cell extracts of PRMT6 transfected 293T cells). Staining of the PRMT6 band in immunoblotting is specifically inhibited by the PRMT6 immunizing peptide. Predicted molecular weight: 38 kDa. Not tested in other applications. Optimal dilutions/concentrations should be determined by the end user.

Type: Primary
Antigen: PRMT6
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR