Anti-PRMT2 Rabbit Polyclonal Antibody

Supplier: GeneTex
GTX23763
89361-380EA 577.28 USD
89361-380
Anti-PRMT2 Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal to PRMT2 - ChIP Grade

ChIP: Use 10?l for 10^6 cells. ELISA: Use at a dilution of 1/1000. WB: Use at a dilution of 1/100 - 1/500. Predicted molecular weight: 48 kDa. Not tested in other applications. Optimal dilutions/concentrations should be determined by the end user.

Type: Primary
Antigen: PRMT2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR