Anti-PPARA Rabbit Polyclonal Antibody

Supplier: GeneTex
GTX44509
89363-542EA 597.66 USD
89363-542
Anti-PPARA Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal to PPAR alpha

Recommended dilutions: WB: Use at a concentration of 3 - 6 ?g/ml. Detects a band of approximately 52 kDa (predicted molecular weight: 52 kDa). Not yet tested in other applications. Optimal dilutions/concentrations should be determined by the end user.

Type: Primary
Antigen: PPARA
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR