Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-PNRA Mouse Monoclonal Antibody [clone: B461]

Supplier: Genetex

GTX72167
89352-956EA 325.28 USD InStock
89352-956
Anti-PNRA Mouse Monoclonal Antibody [clone: B461]
Antibodies
Monoclonal Mouse Anti-Human Renal cell carcinoma (Proximal Nephrogenic Renal Antigen)

Type: Primary
Antigen: PNRA
Clonality: Monoclonal
Clone: B461
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty