Anti-PMS2 Rabbit Polyclonal Antibody

Supplier: GeneTex
GTX113804
89357-744EA 557.66 USD
89357-744
Anti-PMS2 Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody to PMS2 (PMS2 postmeiotic segregation increased 2 (S. cerevisiae))

Suggested starting dilutions are as follows: WB: 1:500-1:10000, IP: 1:100-1:500. Not yet tested in other applications. Optimal working dilutions should be determined experimentally by the end user.

Type: Primary
Antigen: PMS2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR