Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Due to system maintenance, Order Status will not be shown from 8:00am – 10:00am CEST Sunday.

Anti-PMP22 Mouse Monoclonal Antibody [clone: 20-14]

Supplier: Genetex

GTX72231
89352-978EA 358.9 USD InStock
89352-978
Anti-PMP22 Mouse Monoclonal Antibody [clone: 20-14]
Antibodies
Mouse monoclonal [20-14] to PMP-22 (Peripheral Myelin Protein)

For WB: Use at an assay dependent dilution. Not tested in other applications. Optimal dilutions/concentrations should be determined by the end user.

Type: Primary
Antigen: PMP22
Clonality: Monoclonal
Clone: 20-14
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgM
Reactivity: Human

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty