Anti-PLTP Rabbit Polyclonal Antibody

Supplier: GeneTex
GTX30230
89362-894EA 587.29 USD
89362-894
Anti-PLTP Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal to PLTP

For WB: Use at a dilution of 1:500 - 1:1,000. May be used in western analysis where a band is seen at ~55kDa representing PLTP. This antibody does not exhibit neutralizing activity. Not tested in other applications. Optimal dilutions/concentrations should be determined by the researcher.

Type: Primary
Antigen: PLTP
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR