Anti-PLK1 Mouse Monoclonal Antibody [clone: Mixed clones]

Supplier: GeneTex
GTX14210
89359-960EA 591 USD
89359-960
Anti-PLK1 Mouse Monoclonal Antibody [clone: Mixed clones]
Antibodies
Polo-like kinases are a family of serine/threonine protein kinases, named after the prototypic founding member of the family, the polo gene product of Drosophila melanogaster. The polo kinase was originally identified in mutants that display abnormal mitotic spindle organization. Subsequently, potential homologues of Drosophila polo have been identified in yeasts (Cdc5p in Saccharomyces cerevisiae; plo1+ in Schizosaccharmoyces pombe) and in mammals (polo-like kinase 1; Plk1). Genetic and biochemical studies suggest that polo, Cdc5p and plo1+ may be required for mitotic spindle organization and, possibly, for cytokinesis. Likewise, the patterns of expression, activity and subcellular localization of Plk1 suggest that Plk12 may function during mitosis in spindle assembly and function. Recent studies have demonstrated that mammalian Plks regulate the function of the Golgi complex, a cellular organelle closely associated with the centrosome and also have microtubule organization activity. Furthermore, deregulated expression of human PLK1 and PLK3 is strongly correlated with the development of many types of malignancies, and ectopic expression of kinase-active Plk3 or Plk1 dominant negative protein leads to rapid cell death.

ELISA: Use at a concentration of 0.1 - 1 ?g/ml. IF: Use at a concentration of 2 - 5 ?g/ml. IP: Use at a concentration of 2 - 5 ?g/ml. WB: Use at a concentration of 1 - 2 ?g/ml. Detects a band of approximately 68 kDa. Not tested in other applications. Optimal dilutions/concentrations should be determined by the end user.

Type: Primary
Antigen: PLK1
Clonality: Monoclonal
Clone: Mixed clones
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR