Anti-PLEK Goat Polyclonal Antibody

Supplier: GeneTex
GTX17020
89360-258EA 578.07 USD
89360-258
Anti-PLEK Goat Polyclonal Antibody
Antibodies
Pleckstrin is the major protein kinase C substrate of platelets however its exact function is not known.

Approx 45kDa band observed in Human Heart and Peripheral Blood Mononucleocyte lysates (calculated MW of 40.1kDa according to NP_002655.2). Recommended for use at 0.05-1µg/ml.

Type: Primary
Antigen: PLEK
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR