Anti-Platelet Activating Receptor H963 Rabbit Polyclonal Antibody

Supplier: GeneTex
GTX13378
89359-652EA 481.52 USD
89359-652
Anti-Platelet Activating Receptor H963 Rabbit Polyclonal Antibody
Antibodies
H963 is an orphan receptor that is a member of the Platelet Activating Factor Receptor subfamily. ESTs have been isolated from normal human blood (activated), liver, and testis libraries, and from a human uterine adenocarcinoma library.

Type: Primary
Antigen: Platelet Activating Receptor H963
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR