Anti-PKD2 Rabbit Polyclonal Antibody

Supplier: GeneTex
GTX70414
89364-966EA 600.29 USD
89364-966
Anti-PKD2 Rabbit Polyclonal Antibody
Antibodies
Rabbit Polyclonal to Protein Kinase D2 ( PKD2 )

Western blot - Use at 1:500 to 1:1,000 dilution. However, the investigator should determine the optimal dilution for a specific application such as immunohistochemistry.

Type: Primary
Antigen: PKD2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: 0
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR