Anti-PRKACB Rabbit Polyclonal Antibody

Supplier: GeneTex
GTX25816
89361-748EA 582.56 USD
89361-748
Anti-PRKACB Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal to PKA beta ( catalytic subunit ) ( phospho S338 )

WB: Use at a dilution of 1/1000. Detects a band of approximately 42 kDa. Not tested in other applications. Optimal dilutions/concentrations should be determined by the end user.

Type: Primary
Antigen: PRKACB
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Pig, Bovine
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR