Anti-PIP2 Mouse Monoclonal Antibody

Supplier: GeneTex
GTX22335
89361-228EA 591 USD
89361-228
Anti-PIP2 Mouse Monoclonal Antibody
Antibodies
Phosphatidylinositol 4,5-biphosphate (PIP2) has been implicated in a variety of cellular processes, including synaptic vesicle recycling.

Not tested in other applications. Working dilutions are best determined by the end user.

Type: Primary
Antigen: PIP2
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: 0
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR