Anti-IgM Goat Polyclonal Antibody

GTX77133
89353-508EA 154.53 USD
89353-508
Anti-IgM Goat Polyclonal Antibody
Goats were immunized with Pig IgM from normal pig serum. After multiple immunizations in Freund's adjuvant serum was collected. The resulting antiserum was solid phase absorbed and the anti-IgM was immunoaffinity purified off an antigen containing immunoabsorbant. The anti-IgM was then conjugated with horseradish (HRP).

Type: Secondary
Antigen: IgM
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Pig
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR