Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Goat Anti-Pig IgM Polyclonal Antibody

Supplier: Genetex

GTX77134
89353-510
Goat Anti-Pig IgM Polyclonal Antibody
Recommended dilutions for ELISA: Capture: 1:100 A 1:500. Primary: 1:5,000-1:20,000.. Goats were immunized with Pig IgM from normal pig serum. After multiple immunizations in Freund's adjuvant serum was collected. The resulting antiserum was solid phase absorbed and the anti-IgM was immunoaffinity purified off an antigen containing immunoabsorbant

Type: Secondary
Antigen: IgM
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Pig

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty