Anti-PGC1A Goat Polyclonal Antibody

Supplier: GeneTex
GTX89046
89366-972EA 578.23 USD
89366-972
Anti-PGC1A Goat Polyclonal Antibody
Antibodies
Goat polyclonal antibody to PGC1 alpha

Approx 100kDa band observed in Rat Stomach lysates (calculated MW of 91.0kDa according to NP_037393.1. Recommended concentration: 1-3µg/ml.

Type: Primary
Antigen: PGC1A
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Dog
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR