Anti-PERP Rabbit Polyclonal Antibody

Supplier: Genetex

GTX23945
89361-404EA 357.99 USD
89361-404
Anti-PERP Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal to PERP

WB: Use at a dilution of 1/500 - 1/1000. By Western blot, this antibody detects a band of ~23 kDa, which corresponds to the predicted molecular weight of PERP. Not tested in other applications. Optimal dilutions/concentrations should be determined by the end user.

Type: Primary
Antigen: PERP
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR