Anti-PDE8A Rabbit Polyclonal Antibody

Supplier: GeneTex
GTX14618
89367-354EA 585.03 USD
89367-354
Anti-PDE8A Rabbit Polyclonal Antibody
Antibodies
One of the many phosphodiesterases that compartmentalized and hydrolyze cAMP into AMP is phosphodiesterase type 8. The cAMP-specific phosphodiesterase type 8 (PDE8) family is comprised of 2 genes (PDE8A and PDE8B) each with multiple splice variants generated by RNA splicing and use of alternate initiation sites. PDE8 family is a high affinity cAMP-specific, IBMX sensitive PDE. The PDE8 has a significant conserved region of about 270 amino acids common to all PDEs at the carboxy terminal apparently serves as the catalytic domain. The amino-terminal region of this protein is divergent and presumably accounts for the distinctive and regulatory properties unique to the individual PDE families. PDE8A protein showed significant homology to other cAMP-dependent PDEs (23%) within the catalytic domain. PDE8A is widely expressed in various tissues in contrast to PDE8B that is exclusively expressed in thyroid gland. The PDE8A transcripts are found in brain, pancreas, placenta, thyroid, spleen, trachea, prostate, and uterus.

Type: Primary
Antigen: PDE8A
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR