Anti-RUVBL2 Rabbit Polyclonal Antibody

Supplier: Genetex

GTX102037
89349-420EA 427.1 USD
89349-420
Anti-RUVBL2 Rabbit Polyclonal Antibody
Antibodies
Rabbit Polyclonal antibody to RUVBL2 (RuvB-like 2 (E. coli))

Suggested starting dilutions are as follows: WB: 1:1000-1:10000, IHC-P: 1:100-1:1000. Not yet tested in other applications. Optimal working dilutions should be determined experimentally by the end user.

Type: Primary
Antigen: RUVBL2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR