Anti-Rubella Virus Core Antigen Mouse Monoclonal Antibody [clone: 3D2]

Supplier: GeneTex
GTX10013
89350-568EA 382.65 USD
89350-568
Anti-Rubella Virus Core Antigen Mouse Monoclonal Antibody [clone: 3D2]
Antibodies
Mouse monoclonal [3D2] to Rubella Virus core antigen

ELISA: Use at an assay dependent dilution. WB: Use at an assay dependent dilution. Not tested in other applications. Optimal dilutions/concentrations should be determined by the end user.

Type: Primary
Antigen:
Clonality: Monoclonal
Clone: 3D2
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Rubella virus
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR