PSD-95/DLG/ZO-1 (PDZ) domain-containing proteins play a central role in synaptic membrane protein localization. The protein interacting with protein kinase C (PICK 1) is a synaptic PDZ domain protein that also contains a coiled-coil and acidic domain. Studies indicate that PICK 1 functions as a targeting and transport protein and has been shown to interact with protein kinase C alpha (PKC alpha), AMPA-type glutamate receptors, and several other membrane receptors via its PDZ domain. The interaction of PICK 1 with PKC alpha is highly dependent on the activation of the kinase. It appears that PICK 1 directs the activated form of PKC alpha to membrane anchored GluR2. Once phosphorylated by PKC alpha, GluR2 is released from the synaptic anchor proteins. Once released, GluR2 is transported from the synaptic membrane in a PICK 1-dependent manner.
WB: Use at a dilution of 1/500 - 1/1000. Predicted molecular weight: 50 kDa. Not tested in other applications. Optimal dilutions/concentrations should be determined by the end user.
Type: Primary
Antigen: PICK1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse