Anti-PEN2 Rabbit Polyclonal Antibody

Supplier: GeneTex
GTX85064
89366-810EA 608.71 USD
89366-810
Anti-PEN2 Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody to PEN2

Type: Primary
Antigen: PEN2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR