Anti-PDE10A Mouse Monoclonal Antibody [clone: 1C9]

Supplier: GeneTex
GTX83933
89366-782EA 705.1 USD
89366-782
Anti-PDE10A Mouse Monoclonal Antibody [clone: 1C9]
Antibodies
Mouse monoclonal antibody [1C9] to PDE10A

Recommended Starting Dilutions:WB 1:500 IHC 1:50 Optimal working dilutions should be determined experimentally by the end user.

Type: Primary
Antigen: PDE10A
Clonality: Monoclonal
Clone: 1C9
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR