Anti-PC4 Rabbit Polyclonal Antibody

Supplier: GeneTex
GTX77897
89366-398EA 619.97 USD
89366-398
Anti-PC4 Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal to PC4

Recommended Starting Dilutions: For IHC-P: Use at a concentration of 4-8 ?g/ml. For WB: Use at a concentration of 0.5-1.0 ug/ml in 5% skim milk/PBS buffer. Not tested in other applications. Optimal dilutions should be determined by the researcher.

Type: Primary
Antigen: PC4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Dog
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR