Anti-PARP2 Goat Polyclonal Antibody

Supplier: Genetex

GTX89417
89366-986EA 412.14 USD
89366-986
Anti-PARP2 Goat Polyclonal Antibody
Antibodies
Goat polyclonal antibody to PARP2

Approx 70kDa band observed in Human Spleen lysates (calculated MW of 60.8kDa according to NP_005475.1. Recommended concentration: 1-3µg/ml.

Type: Primary
Antigen: PARP2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Dog
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR