Anti-PPY Goat Polyclonal Antibody

Supplier: GeneTex
GTX89539
89366-994EA 581.69 USD
89366-994
Anti-PPY Goat Polyclonal Antibody
Antibodies
Goat polyclonal antibody to Pancreatic Polypeptide / PPY

Preliminary experiments gave an approx 35-38kDa band in human pancreas lysates after 1µg/ml antibody staining. Please note that currently we cannot find an explanation in the literature for the band we observe given the calculated size of 10.5kDa according to NP_002713.1. The 38kDa band was successfully blocked by incubation with the immunizing peptide. In paraffin embedded human pancreas, shows strong cytoplasmic staining of rare cells located at the periphery of islets of Langerhans. Recommended concentration, 3µg/ml.

Type: Primary
Antigen: PPY
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR