Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-PAN Synuclein Rabbit Polyclonal Antibody

Supplier: Genetex

GTX15534
89351-160EA 358.79 USD InStock
89351-160
Anti-PAN Synuclein Rabbit Polyclonal Antibody
Antibodies
Synucleins are small, soluble proteins expressed primarily in neural tissue and in certain tumors. The family includes three known proteins: alpha, beta and gamma synucleins. The alpha- and beta synuclein proteins are found primarily in brain tissue, where they are seen mainly in presynaptic terminals. The gamma synuclein protein is found primarily in the peripheral nervous system and retina but its expression in breast tumors is a marker for tumor progression.

Type: Primary
Antigen: PAN Synuclein
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty