Anti-TP73 Mouse Monoclonal Antibody [clone: 5B429]

Supplier: GeneTex

GTX46123
89363-594EA 427.16 USD
89363-594
Anti-TP73 Mouse Monoclonal Antibody [clone: 5B429]
Antibodies
Mouse monoclonal antibody [5B429] to p73

IF: Use at an assay dependent dilution. IP: Use at an assay dependent dilution. WB: 1/500. Predicted molecular weight: 70 kDa. IF: Use at an assay dependent dilution. Not yet tested in other applications. Optimal dilutions/concentrations should be determined by the end user.

Type: Primary
Antigen: TP73
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR