Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-P2Y12 Rabbit Polyclonal Antibody

Supplier: Genetex

GTX71193
89365-210EA 381.65 USD InStock
89365-210
Anti-P2Y12 Rabbit Polyclonal Antibody
Antibodies
P2Y12 is a Purinergic Receptor essential for the aggregation of blood platelets. There is evidence that mutations in P2Y12 cause a bleeding disorder, suggesting that knowledge of the P2Y12 receptor could advance the development of antiplatelet agents to treat cardiovascular diseases. Expression of P2Y12 has been reported in platelets, spinal cord, and many brain regions. ESTs for P2Y12 have been isolated from B-cell/lung/testis, brain, embryo, prostate, eye, kidney carcinoma, and colon carcinoma libraries. Cognate

Type: Primary
Antigen: P2Y12
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty