Anti-CDNK1A Mouse Monoclonal Antibody [clone: WA-1]

Supplier: GeneTex
GTX27903
89362-682EA 546.71 USD
89362-682
Anti-CDNK1A Mouse Monoclonal Antibody [clone: WA-1]
Antibodies
Mouse monoclonal antibody [WA - 1] to p21 Cip1

ELISA: Use at an assay dependant concentration. WB: Use at an assay dependant concentration. Not tested in other applications. Optimal dilutions/concentrations should be determined by the end user.

Type: Primary
Antigen: CDNK1A
Clonality: Monoclonal
Clone: 0
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR