Anti-OXTR Goat Polyclonal Antibody

Supplier: GeneTex
GTX31093
89363-028EA 322.27 USD
89363-028
Anti-OXTR Goat Polyclonal Antibody
Antibodies
Goat polyclonal antibody to Oxytocin receptor

Optimal concentration should be determined experimentally by the end user.

Type: Primary
Antigen: OXTR
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR