Anti-TFR1 Mouse Monoclonal Antibody [clone: MEM-75]

Supplier: GeneTex
GTX29179
89362-794EA 617.47 USD
89362-794
Anti-TFR1 Mouse Monoclonal Antibody [clone: MEM-75]
Antibodies
Mouse monoclonal antibody [MEM-75] to CD71

FACS: Use at an assay dependant concentration. IP: Use at an assay dependant concentration. Not tested in other applications. Optimal dilutions/concentrations should be determined by the end user.

Type: Primary
Antigen: TFR1
Clonality: Monoclonal
Clone: MEM-75
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR