Anti-TF Mouse Monoclonal Antibody [clone: 8B9]

Supplier: GeneTex
GTX10212
89350-700EA 696.18 USD
89350-700
Anti-TF Mouse Monoclonal Antibody [clone: 8B9]
Antibodies
Mouse monoclonal [8B9] to Transferrin

ELISA: Use at an assay dependent dilution. Will detect transferrin in serum by sandwich ELISA as an antibody pair. Use GTX10212 as the coating agent with either GTX10208 or GTX10209 for detection. Will detect transferrin-transferrin receptor complex when used as an antibody pair with an anti-transferrin receptor antibody. Not tested in other applications. Optimal dilutions/concentrations should be determined by the end user.

Type: Primary
Antigen: TF
Clonality: Monoclonal
Clone: 8B9
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR