Anti-TF Chicken Polyclonal Antibody

Supplier: GeneTex
GTX82014
89368-034EA 617.31 USD
89368-034
Anti-TF Chicken Polyclonal Antibody
Antibodies
Chicken polyclonal to Serotransferrin

ELISA (Only tested on recombinant protein): Affinity-Purified anti-Transferrin IgY as primary antibody (0.1 ug/ml). and Rabbit anti-IgY-HRP as 2nd antibody. WB (only tested on plasma transferrin protein): Use at a dilution of 1:2000. Colorimetric method for signal development.

Type: Primary
Antigen: TF
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Chicken
Isotype: IgY
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR