Anti-RANBP16 Goat Polyclonal Antibody

Supplier: GeneTex
GTX26041
89361-810EA 607.87 USD
89361-810
Anti-RANBP16 Goat Polyclonal Antibody
Antibodies
Goat polyclonal antibody to RANBP16 / Exportin 7

Approx 110kDa band observed in Human Testis and Hela lysates (predicted size of approx. 124kDa according to NP_055839). Recommended for use at 0.5-1µg/ml This antibody is expected to recognise all the reported isoforms (NP_001093631.1; NP_055839; NP_001093632.1).

Type: Primary
Antigen: RANBP16
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Bovine
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR