Anti-PPM1A Mouse Monoclonal Antibody [clone: p6c7]

Supplier: GeneTex
GTX14824
89360-068EA 560.3 USD
89360-068
Anti-PPM1A Mouse Monoclonal Antibody [clone: p6c7]
Antibodies
PPM1A is a Mn2+- or Mg2+-dependent protein serine/threonine phosphatase that inhibits the human stress-responsive p38 and JNK MAPK pathways and regulates cellular stress response in eukaryotes. The PPM (metal-dependent protein phosphatase) family of Ser/Thr protein phosphatases have recently been shown to down-regulate the stress response pathways in eukaryotes. Within the stress pathway, key signaling kinases, which are activated by protein phosphorylation, have been proposed as the in vivo substrates of PP2C, the prototypical member of the PPM family.

ELISA: Use at an assay dependent dilution. WB: Use at a dilution of 1/250 - 1/1000. Predicted molecular weight: 42 kDa. Not tested in other applications. Optimal dilutions/concentrations should be determined by the end user.

Type: Primary
Antigen: PPM1A
Clonality: Monoclonal
Clone: p6c7
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR