Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-OVA Mouse Monoclonal Antibody

Supplier: Genetex

GTX83272
89353-896EA 367.81 USD InStock
89353-896
Anti-OVA Mouse Monoclonal Antibody
Antibodies
Ovalbumin (OVA) is used in variety of laboratories products which contain OVA as a carrier protein. To raise antibodies against small non-immunogenic molecules,called haptens,it is often necessary to couple them to carrier protein,such as OVA.This antibody is a good reagent that may be used for the development of detection assay in ELISA or western blot to test the remaning OVA for clinical products in Vivo or research products In Vitro.

Type: Primary
Antigen: OVA
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: 0

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty