Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-OPRM1 Rabbit Polyclonal Antibody

Supplier: Genetex

GTX71045
89365-160EA 381.65 USD InStock
89365-160
Anti-OPRM1 Rabbit Polyclonal Antibody
Antibodies
OPRM1, an Opioid Receptor, mediates the effects of opioids such as morphine, heroin, fentanyl, methadone, and beta-endorphin. It is involved in the control of addiction, pain, and other mood-altering disorders. Expression of OPRM1 has been reported mostly in brain and spinal cord.

Type: Primary
Antigen: OPRM1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty