Anti-Mouse HRP Antibody

Supplier: GeneTex
GTX83400
89366-770EA 304.69 USD
89366-770
Anti-Mouse HRP Antibody
Antibodies
Immunohistochemistry (IHC) is the localization of antigens in tissue sections/cells using antibodies as specific reagents. Visualization of antibody signal is often significantly improved using the biotin-avidin amplification system. This system, however, adds significant time to experimental protocols and results in higher background due to the presence of endogenous biotin in tissue.

GeneTex’s One-Step anti-mouse and rat IgG (H+L) uses the power of next-generation Polymer technology to provide dramatic signal amplification without the need for biotin. This reagent has been shown to provide increased sensitivity and detection, while reducing the incubation time and required quantity of primary antibody. This reagent can be used to enhance staining of membranes, cytoplasmic and nuclear antigens. It provides the user with a rapid and easy to use IHC detection system.
<b>Features and Benefits</b>

‧ Lower Amplification without biotin avoids background from endogenous biotin
‧ Higher sensitivity than standard IHC detection
‧ Faster: Staining time reduced to 15 minutes!
‧ Compatible with either manual or automated systems
‧ Better results on difficult-to-work antibodies and nuclear staining
‧ Save cost due to further dilution of primary antibody

Type: Secondary
Antigen: HRP
Clonality:
Clone:
Conjugation: Unconjugated
Epitope:
Host:
Isotype:
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR