Anti-POU2F2 Rabbit Polyclonal Antibody

GTX15114
89350-954EA 376.73 USD
89350-954
Anti-POU2F2 Rabbit Polyclonal Antibody
Oct2 is a transcription factor that specifically binds to the octamer motif (5'-ATTTGCAT-3'). Oct2 regulates transcription in a number of tissues in addition to activating immunoglobulin gene expression. It also modulates transcription transactivation by NR3C1, AR and PGR. Oct2 is B-cell specific, belongs to the POU transcription factor family class-2 subfamily and contains 1 homeobox domain.

IHC-P: Use at a dilution of 1/50. Perform heat mediated antigen retrieval before commencing with IHC staining protocol. IP: Use at a dilution of 2 ?g/mg protein lysate. WB: Use at a concentration of 1 - 2 ?g/ml. Predicted molecular weight: 51 kDa. Not tested in other applications. Optimal dilutions/concentrations should be determined by the end user.

Type: Primary
Antigen: POU2F2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR