Anti-NURR Rabbit Polyclonal Antibody

Supplier: GeneTex
GTX114045
89357-846EA 582.76 USD
89357-846
Anti-NURR Rabbit Polyclonal Antibody
Antibodies
Rabbit Polyclonal antibody to Nurr1 (nuclear receptor subfamily 4, group A, member 2)

Suggested starting dilutions are as follows: ICC/IF: 1:100-1:1000. Not yet tested in other applications. Optimal working dilutions should be determined experimentally by the end user.

Type: Primary
Antigen: NURR
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR