Anti-NUMA Rabbit Polyclonal Antibody

Supplier: GeneTex
GTX30688
89367-690EA 612.52 USD
89367-690
Anti-NUMA Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody to NuMA (Nuclear Mitotic Apparatus)

This antibody may be used for Western blotting, immunoprecipitation, and immunofluorescence (frozen sections tested). A band at 220 kDa can be detected by Western blot. Other applications have not been tested.Suggested working dilutions: *Western Blot - 1:1,000 Immunofluorescence - 1:500 *The investigator should determine the optimal working dilution for a specific application.

Type: Primary
Antigen: NUMA
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR