Anti-ENO2 Mouse Monoclonal Antibody [clone: 500]

GTX71959
89352-916EA 341.32 USD
89352-916
Anti-ENO2 Mouse Monoclonal Antibody [clone: 500]
Mouse Monoclonal to Human Neuron - Specific Enolase

Prolonged fixation in buffered formalin can destroy the epitope. The antibody may be used at a dilution of 1:50 to 1:100 with AutoProbe III (GTX28620). Dilute according to the particular application being used. In general, the 0.05M Borate pH 8.0 containing 0.15M Sodium Chloride, 0.05% Sodium Azide, is a good diluent to use with most antibodies. Not tested in other applications. Optimal dilutions/concentrations should be determined by the researcher.

Type: Primary
Antigen: ENO2
Clonality: Monoclonal
Clone: 500
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR